accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3SH

CAS RN 25697-56-7

IUPAC Name: (3-methylphenyl)methanethiol
Synonyms: (3-methylphenyl)methanethiol
Molecular Weight (g/mol): 138.23
Molecular Formula: C8H10S
InChi Key: PNAZUQUHTIOEHF-UHFFFAOYSA-N
SMILES: CC1=CC(CS)=CC=C1
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)