accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 25702-80-1

Rn

CAS RN 25702-80-1

IUPAC Name: chloroethene; prop-2-enoic acid
Synonyms: acrylic acid; vinyl chloride
Molecular Weight (g/mol): 134.56
Molecular Formula: C5H7ClO2
InChi Key: SQNNHEYXAJPPKH-UHFFFAOYSA-N
SMILES: ClC=C.OC(=O)C=C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results