accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

∗∗SOOONan

CAS RN 25704-18-1

IUPAC Name: 4-ethenylbenzene-1-sulfonic acid
Synonyms: tolevamer
Molecular Weight (g/mol): 184.21
Molecular Formula: C8H8O3S
InChi Key: MAGFQRLKWCCTQJ-UHFFFAOYSA-N
SMILES: OS(=O)(=O)C1=CC=C(C=C)C=C1
Quantity 
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (9)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (6)