accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 25706-29-0

IUPAC Name: 2-(methylsulfanyl)-1,3-benzothiazol-6-amine
Synonyms: 2-(methylsulfanyl)-1,3-benzothiazol-6-amine
Molecular Weight (g/mol): 196.29
Molecular Formula: C8H8N2S2
InChi Key: GYCJZSAIEMSFAV-UHFFFAOYSA-N
SMILES: CSC1=NC2=CC=C(N)C=C2S1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results