accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 257933-82-7

CH3ONNHNFClNHONH3CCH3(E)

CAS RN 257933-82-7

IUPAC Name: (2E)-N-{4-[(3-chloro-4-fluorophenyl)amino]-3-cyano-7-ethoxyquinolin-6-yl}-4-(dimethylamino)but-2-enamide
Synonyms: pelitinib
Molecular Weight (g/mol): 467.93
Molecular Formula: C24H23ClFN5O2
InChi Key: WVUNYSQLFKLYNI-AATRIKPKSA-N
SMILES: CCOC1=C(NC(=O)\C=C\CN(C)C)C=C2C(NC3=CC=C(F)C(Cl)=C3)=C(C=NC2=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results