accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

∗N∗OMen

CAS RN 25805-17-8

IUPAC Name: 2-ethyl-4,5-dihydro-1,3-oxazole
Synonyms: polyethyloxazoline
Molecular Weight (g/mol): 99.13
Molecular Formula: C5H9NO
InChi Key: NYEZZYQZRQDLEH-UHFFFAOYSA-N
SMILES: CCC1=NCCO1
Quantity 
  • (3)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (3)
  • (3)
  • (3)