accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONONNNH(E/Z)

CAS RN 2581-69-3

IUPAC Name: 4-[2-(4-nitrophenyl)diazen-1-yl]-N-phenylaniline
Synonyms: disperse orange 1
Molecular Weight (g/mol): 318.34
Molecular Formula: C18H14N4O2
InChi Key: YFVXLROHJBSEDW-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C=C1)N=NC1=CC=C(NC2=CC=CC=C2)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results