accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2NH2NH2NOHOHO

CAS RN 2582-30-1

IUPAC Name: N''-aminoguanidine; carbonic acid
Synonyms: amino-guanidine; carbonic acid
Molecular Weight (g/mol): 136.11
Molecular Formula: C2H8N4O3
InChi Key: OTXHZHQQWQTQMW-UHFFFAOYSA-N
SMILES: OC(O)=O.NN=C(N)N
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (6)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)