accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 25895-60-7

NBHHHNa

CAS RN 25895-60-7

IUPAC Name: boron(3+) sodium iminomethanide trihydride
Synonyms: sodium cyanoborohydride
Molecular Weight (g/mol): 62.84
Molecular Formula: CH3BNNa
InChi Key: IVGYREMSCQFSBM-UHFFFAOYSA-N
SMILES: [H-].[H-].[H-].[B+3].[Na+].[C-]#N
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (6)
  • (2)
  • (3)