accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 259-79-0

IUPAC Name: biphenylene
Synonyms: diphenylene cyclobutadibenzene dibenzocyclobutadiene 1,1'-biphenylene
Molecular Weight (g/mol): 152.2
Molecular Formula: C12H8
InChi Key: IFVTZJHWGZSXFD-UHFFFAOYSA-N
SMILES: C1=CC=C2C(=C1)C1=CC=CC=C21

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results