accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ON

CAS RN 2591-86-8

IUPAC Name: piperidine-1-carbaldehyde
Synonyms: N-formylpiperidine
Molecular Weight (g/mol): 113.16
Molecular Formula: C6H11NO
InChi Key: FEWLNYSYJNLUOO-UHFFFAOYSA-N
SMILES: O=CN1CCCCC1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results