accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNHHNNOO

CAS RN 25948-11-2

IUPAC Name: 5-nitro-2-(propylamino)pyridin-1-ium
Synonyms: 5-nitro-2-(propylamino)pyridin-1-ium
Molecular Weight (g/mol): 182.2
Molecular Formula: C8H12N3O2
InChi Key: KZWODZHPEMBNTR-UHFFFAOYSA-O
SMILES: CCCNC1=CC=C(C=[NH+]1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results