accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 25962-68-9

NSNHNOCl

CAS RN 25962-68-9

IUPAC Name: 2-chloro-N-(3-phenyl-1,2,4-thiadiazol-5-yl)acetamide
Synonyms: 2-chloro-N-(3-phenyl-1,2,4-thiadiazol-5-yl)acetamide
Molecular Weight (g/mol): 253.7
Molecular Formula: C10H8ClN3OS
InChi Key: DSELUSKAJCJQHM-UHFFFAOYSA-N
SMILES: ClCC(=O)NC1=NC(=NS1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results