CAS RN 259793-88-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
6-Bromo-3-hydroxypyrazine-2-carboxamide, TRC
CAS: 259793-88-9 Molecular Formula: C5 H4 Br N3 O2 Molecular Weight (g/mol): 218.008 Synonym: 2-Pyrazinecarboxamide, 6-bromo-3,4-dihydro-3-oxo- (ACI),6-Bromo-3,4-dihydro-3-oxo-2-pyrazinecarboxamide (ACI),Pyrazinecarboxamide, 6-bromo-3,4-dihydro-3-oxo- (9CI),6-Bromo-3-hydroxypyrazine-2-carboxamide,Br-T-705 IUPAC Name: 5-bromo-2-oxo-1H-pyrazine-3-carboxamide SMILES: NC(=O)C1=NC(=CNC1=O)Br