accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NNNOOH

CAS RN 259807-47-1

IUPAC Name: 2-(pyridin-4-yl)pyrimidine-5-carboxylic acid
Synonyms: 2-(pyridin-4-yl)pyrimidine-5-carboxylic acid
Molecular Weight (g/mol): 201.19
Molecular Formula: C10H7N3O2
InChi Key: GTKJGOOMSODGOU-UHFFFAOYSA-N
SMILES: OC(=O)C1=CN=C(N=C1)C1=CC=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results