CAS RN 25984-63-8
CAS RN 25984-63-8
IUPAC Name:
4-[amino(sulfanyl)methylidene]cyclohexa-2,5-dien-1-one
Synonyms:
4-[amino(sulfanyl)methylidene]cyclohexa-2,5-dien-1-one
Molecular Weight (g/mol):
153.2
Molecular Formula:
C7H7NOS
InChi Key:
DOCQBKMMNPJLOR-UHFFFAOYSA-N
SMILES:
NC(S)=C1C=CC(=O)C=C1
1
–
1
of
1
results
1
4-Hydroxythiobenzamide, 98%
CAS: 25984-63-8 Molecular Formula: C7H7NOS Molecular Weight (g/mol): 153.20 MDL Number: MFCD04973332 InChI Key: DOCQBKMMNPJLOR-UHFFFAOYSA-N Synonym: 4-hydroxythiobenzamide,4-hydroxybenzothioamide,4-hydroxy thiobenzamide,4-hydroxybenzene-1-carbothioamide,4-hydroxybenzenecarbothioamide,benzenecarbothioamide, 4-hydroxy,4-hydroxy-thiobenzamide,4-aminothioxomethyl phenol,4-amino sulfanyl methylidene cyclohexa-2,5-dien-1-one,p-hydroxythiobenzamide PubChem CID: 5706487 IUPAC Name: 4-[amino(sulfanyl)methylidene]cyclohexa-2,5-dien-1-one SMILES: NC(S)=C1C=CC(=O)C=C1