accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

N

CAS RN 260-94-6

IUPAC Name: acridine
Synonyms: acridine dibenzo(b,e)pyridine 2,3-benzoquinoline 2,3,5,6-dibenzopyridine akridin benzo(b)quinoline acrydine
Molecular Weight (g/mol): 179.22
Molecular Formula: C13H9N
InChi Key: DZBUGLKDJFMEHC-UHFFFAOYSA-N
SMILES: C1=CC2=CC3=CC=CC=C3N=C2C=C1
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results