accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClClN

CAS RN 2601-89-0

IUPAC Name: 2,3-dichloropropanenitrile
Synonyms: propanenitrile, 2,3-dichloro-
Molecular Weight (g/mol): 123.96
Molecular Formula: C3H3Cl2N
InChi Key: RJJDLPQZNANQDQ-UHFFFAOYNA-N
SMILES: ClCC(Cl)C#N
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)