accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 26042-64-8

FFFFFFAgSb

CAS RN 26042-64-8

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 343.62
Molecular Formula: AgF6Sb
InChi Key: VWZIPIHZVCRLOX-UHFFFAOYSA-N
SMILES: [F].[F].[F].[F].[F].[F].[Ag+].[Sb]
Molecular Weight (g/mol) 
  • (2)
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)