accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 26060-30-0

IUPAC Name: 4-nitrobenzene-1-carbothioamide
Synonyms: 4-nitrobenzenecarbothioamide
Molecular Weight (g/mol): 182.2
Molecular Formula: C7H6N2O2S
InChi Key: MDBQPBMIZPCKAJ-UHFFFAOYSA-N
SMILES: NC(=S)C1=CC=C(C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results