accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OONH2

CAS RN 26093-31-2

IUPAC Name: 7-amino-4-methyl-2H-chromen-2-one
Synonyms: 7-amino-4-methylcoumarin NH2mec coumarin 120
Molecular Weight (g/mol): 175.19
Molecular Formula: C10H9NO2
InChi Key: GLNDAGDHSLMOKX-UHFFFAOYSA-N
SMILES: CC1=CC(=O)OC2=CC(N)=CC=C12
Molecular Weight (g/mol) 
  • (5)
  • (7)
Percent Purity 
  • (1)
  • (11)
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
Grade 
  • (3)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (4)
  • (3)
  • (1)
  • (5)
  • (5)
  • (7)
  • (1)
  • (11)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (4)