accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 26159-34-2

IUPAC Name: sodium (2S)-2-(6-methoxynaphthalen-2-yl)propanoate
Synonyms: naproxen sodium
Molecular Weight (g/mol): 252.25
Molecular Formula: C14H13NaO3
InChi Key: CDBRNDSHEYLDJV-FVGYRXGTSA-M
SMILES: [Na+].COC1=CC=C2C=C(C=CC2=C1)[C@H](C)C([O-])=O
Molecular Weight (g/mol) 
  • (5)
  • (5)
Quantity 
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results