accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 261732-38-1

H3CH3CCH3OONBr

CAS RN 261732-38-1

IUPAC Name: tert-butyl 5-bromo-2,3-dihydro-1H-indole-1-carboxylate
Synonyms: tert-butyl 5-bromo-2,3-dihydroindole-1-carboxylate
Molecular Weight (g/mol): 298.18
Molecular Formula: C13H16BrNO2
InChi Key: UOCVSZYBRMGQOL-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCC2=C1C=CC(Br)=C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results