accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 261763-29-5

H3COOHFF

CAS RN 261763-29-5

IUPAC Name: 2,3-difluoro-4-methoxyphenol
Synonyms: 2,3-difluoro-4-methoxyphenol
Molecular Weight (g/mol): 160.12
Molecular Formula: C7H6F2O2
InChi Key: CKKAZWYLHGNVDZ-UHFFFAOYSA-N
SMILES: COC1=CC=C(O)C(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results