accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3FOCH3F

CAS RN 261763-33-1

IUPAC Name: 1,3-difluoro-2-methoxy-4-methylbenzene
Synonyms: 1,3-difluoro-2-methoxy-4-methylbenzene
Molecular Weight (g/mol): 158.15
Molecular Formula: C8H8F2O
InChi Key: ZOVHZEAOMNTVFB-UHFFFAOYSA-N
SMILES: COC1=C(F)C=CC(C)=C1F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results