accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OHFF

CAS RN 261763-45-5

IUPAC Name: 2,3-difluoro-4-methylphenol
Synonyms: 2,3-difluoro-4-methylphenol
Molecular Weight (g/mol): 144.12
Molecular Formula: C7H6F2O
InChi Key: NVYIUITYVMVHJM-UHFFFAOYSA-N
SMILES: CC1=C(F)C(F)=C(O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results