accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrFFFF

CAS RN 261951-85-3

IUPAC Name: 1-bromo-3-fluoro-2-(trifluoromethyl)benzene
Synonyms: 1-bromo-3-fluoro-2-(trifluoromethyl)benzene
Molecular Weight (g/mol): 243
Molecular Formula: C7H3BrF4
InChi Key: LOXSSZUHCPBDMD-UHFFFAOYSA-N
SMILES: FC1=C(C(Br)=CC=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results