accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HONNNCl

CAS RN 26198-19-6

IUPAC Name: 6-chloro-1H-1,2,3-benzotriazol-1-ol
Synonyms: 6-chloro-1,2,3-benzotriazol-1-ol
Molecular Weight (g/mol): 169.57
Molecular Formula: C6H4ClN3O
InChi Key: TZCYLJGNWDVJRA-UHFFFAOYSA-N
SMILES: ON1N=NC2=CC=C(Cl)C=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results