accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2620-44-2

ONOOO

CAS RN 2620-44-2

IUPAC Name: 5-nitro-2H-1,3-benzodioxole
Synonyms: 5-nitro-2H-1,3-benzodioxole
Molecular Weight (g/mol): 167.12
Molecular Formula: C7H5NO4
InChi Key: SNWQAKNKGGOVMO-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C2OCOC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results