accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2OO

CAS RN 2620-50-0

IUPAC Name: 1-(2H-1,3-benzodioxol-5-yl)methanamine
Synonyms: 1-(2H-1,3-benzodioxol-5-yl)methanamine
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: ZILSBZLQGRBMOR-UHFFFAOYSA-N
SMILES: NCC1=CC=C2OCOC2=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)