accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2623-23-6

H3CCH3(S)(R)CH3(R)OCH3O

CAS RN 2623-23-6

IUPAC Name: (1R,2S,5R)-5-methyl-2-(propan-2-yl)cyclohexyl acetate
Synonyms: menthyl acetate l-menthyl acetate (natural) L-menthyl acetate
Molecular Weight (g/mol): 198.31
Molecular Formula: C12H22O2
InChi Key: XHXUANMFYXWVNG-ADEWGFFLSA-N
SMILES: CC(C)[C@@H]1CC[C@@H](C)C[C@H]1OC(C)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results