accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2626-52-0

H3CNH3CNCS

CAS RN 2626-52-0

IUPAC Name: diethyl(3-isothiocyanatopropyl)amine
Synonyms: diethyl(3-isothiocyanatopropyl)amine
Molecular Weight (g/mol): 172.29
Molecular Formula: C8H16N2S
InChi Key: QPBVKSLJBRCKIF-UHFFFAOYSA-N
SMILES: CCN(CC)CCCN=C=S
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results