accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2ONN(R)O(R)HO(S)HO(R)OHNH2

CAS RN 2627-69-2

IUPAC Name: 5-amino-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1H-imidazole-4-carboxamide
Synonyms: acadesine
Molecular Weight (g/mol): 258.23
Molecular Formula: C9H14N4O5
InChi Key: RTRQQBHATOEIAF-UUOKFMHZSA-N
SMILES: NC(=O)C1=C(N)N(C=N1)[C@@H]1O[C@H](CO)[C@@H](O)[C@H]1O
Percent Purity 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)