accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2627-86-3

IUPAC Name: (1S)-1-phenylethan-1-amine
Synonyms: (S)-α-phenylethylamine L(-)-α-methylbenzylamine (S)-α-methylbenzenemethanamine L-α-methylbenzylamine L-(-)-α-phenylethylamine (-)-α-phenethylamine L-(-)-1-phenylethylamine
Molecular Weight (g/mol): 121.18
Molecular Formula: C8H11N
InChi Key: RQEUFEKYXDPUSK-ZETCQYMHSA-N
SMILES: C[C@H](N)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (7)
  • (6)
Quantity 
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (6)
  • (6)
Percent Purity 
  • (3)
  • (3)
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (7)
  • (6)
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (6)
  • (6)
  • (3)
  • (3)
  • (6)
  • (6)