accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHCH2

CAS RN 2628-17-3

IUPAC Name: 4-ethenylphenol
Synonyms: 4-vinylphenol P-vinylphenol P-vinyl-phenol phenol, P-vinyl-
Molecular Weight (g/mol): 120.15
Molecular Formula: C8H8O
InChi Key: FUGYGGDSWSUORM-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results