accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NNNH2

CAS RN 26286-55-5

IUPAC Name: 2-(2-methyl-1H-imidazol-1-yl)aniline
Synonyms: 2-(2-methylimidazol-1-yl)aniline
Molecular Weight (g/mol): 173.22
Molecular Formula: C10H11N3
InChi Key: LLSZMUSXQITSSJ-UHFFFAOYSA-N
SMILES: CC1=NC=CN1C1=CC=CC=C1N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results