accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClCrOOOHN

CAS RN 26299-14-9

IUPAC Name: hydrogen (trioxochromio)-λ³-chloranidyl pyridine
Synonyms: hydrogen (trioxochromio)-λ³-chloranidyl pyridine
Molecular Weight (g/mol): 215.55
Molecular Formula: C5H6ClCrNO3
InChi Key: BUCMHZVAGRYTNK-UHFFFAOYSA-O
SMILES: [H+].[Cl-][Cr](=O)(=O)=O.C1=CC=NC=C1
Quantity 
  • (3)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results