accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNHOOCH3H3C

CAS RN 2631-40-5

IUPAC Name: 2-(propan-2-yl)phenyl N-methylcarbamate
Synonyms: isoprocarb mipsin caswell no. 512B isoprocarbe mipcine mipcin etrofolan hytox ppc 3 epa pesticide chemical code 512300
Molecular Weight (g/mol): 193.25
Molecular Formula: C11H15NO2
InChi Key: QBSJMKIUCUGGNG-UHFFFAOYSA-N
SMILES: CNC(=O)OC1=CC=CC=C1C(C)C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results