accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COHO

CAS RN 26311-45-5

IUPAC Name: 4-pentylbenzoic acid
Synonyms: benzoic acid, 4-pentyl-
Molecular Weight (g/mol): 192.26
Molecular Formula: C12H16O2
InChi Key: CWYNKKGQJYAHQG-UHFFFAOYSA-N
SMILES: CCCCCC1=CC=C(C=C1)C(O)=O
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results