accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 26340-89-6

IUPAC Name: dihydrogen methyl (2S)-2-amino-5-[(diaminomethylidene)amino]pentanoate dichloride
Synonyms: dihydrogen methyl (2S)-2-amino-5-[(diaminomethylidene)amino]pentanoate dichloride
Molecular Weight (g/mol): 261.15
Molecular Formula: C7H18Cl2N4O2
InChi Key: XQYZOBNLCUAXLF-XRIGFGBMSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].COC(=O)[C@@H](N)CCCN=C(N)N
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)