accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrOO

CAS RN 2635-13-4

IUPAC Name: 5-bromo-2H-1,3-benzodioxole
Synonyms: 5-bromo-2H-1,3-benzodioxole
Molecular Weight (g/mol): 201.02
Molecular Formula: C7H5BrO2
InChi Key: FBOYMIDCHINJKC-UHFFFAOYSA-N
SMILES: BrC1=CC=C2OCOC2=C1
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
  • (2)
  • (2)
  • (4)
  • (2)