accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2637-37-8

IUPAC Name: 1,2-dihydroquinoline-2-thione
Synonyms: 2(1H)-quinolinethione
Molecular Weight (g/mol): 161.22
Molecular Formula: C9H7NS
InChi Key: KXZSVYHFYHTNBI-UHFFFAOYSA-N
SMILES: S=C1NC2=CC=CC=C2C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results