accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 26377-04-8

CH3SnHHOSOHSnHH3CCH3H3COOCH3H3C

CAS RN 26377-04-8

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 680.2
Molecular Formula: C24H58O4SSn2
InChi Key: QFPUJGBQZANZFW-UHFFFAOYSA-N
SMILES: OS(O)(=O)=O.CCCC[SnH](CCCC)CCCC.CCCC[SnH](CCCC)CCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results