accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 26386-88-9

IUPAC Name: {[azido(phenoxy)phosphoryl]oxy}benzene
Synonyms: diphenylphosphoryl azide
Molecular Weight (g/mol): 275.2
Molecular Formula: C12H10N3O3P
InChi Key: SORGEQQSQGNZFI-UHFFFAOYSA-N
SMILES: [N-]=[N+]=NP(=O)(OC1=CC=CC=C1)OC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (6)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (6)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (6)