accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 26388-13-6

IUPAC Name: 1-(2-phenoxyphenyl)ethan-1-one
Synonyms: 1-(2-phenoxyphenyl)ethanone
Molecular Weight (g/mol): 212.25
Molecular Formula: C14H12O2
InChi Key: KPBCVVSDGJBODL-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=CC=C1OC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results