accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 265330-98-1

IUPAC Name: 5-ethoxy-5-oxopentan-1-ide; bromozincylium
Synonyms: 5-ethoxy-5-oxopentan-1-ide; bromozincylium
Molecular Weight (g/mol): 274.46
Molecular Formula: C7H13BrO2Zn
InChi Key: JJSSUZYKRQXNFR-UHFFFAOYSA-M
SMILES: [Zn+]Br.CCOC(=O)CCC[CH2-]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results