accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 26570-48-9

H2COHOOHHO

CAS RN 26570-48-9

IUPAC Name: ethane-1,2-diol; prop-2-enoic acid
Synonyms: acrylic acid; ethylene glycol
Molecular Weight (g/mol): 134.13
Molecular Formula: C5H10O4
InChi Key: FCDZZFLRLVQGEH-UHFFFAOYSA-N
SMILES: OCCO.OC(=O)C=C
Quantity 
  • (3)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (1)
  • (3)
  • (3)
  • (3)