accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOSClOO

CAS RN 26638-43-7

IUPAC Name: methyl 2-(chlorosulfonyl)benzoate
Synonyms: methyl 2-(chlorosulfonyl)benzoate
Molecular Weight (g/mol): 234.65
Molecular Formula: C8H7ClO4S
InChi Key: HUNUAFNLLYVTQD-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC=C1S(Cl)(=O)=O
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)