accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 26663-77-4

CH3OONHN

CAS RN 26663-77-4

IUPAC Name: methyl 1H-1,3-benzodiazole-6-carboxylate
Synonyms: methyl 3H-1,3-benzodiazole-5-carboxylate
Molecular Weight (g/mol): 176.18
Molecular Formula: C9H8N2O2
InChi Key: WJHHIVYNOVTVGY-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C2N=CNC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results