CAS RN 2675-77-6
IUPAC Name:
1,4-dichloro-2,5-dimethoxybenzene
Synonyms:
chloroneb caswell no. 198 nuflo D flo pro D seed protectant demosan Tersan-SP demosan 10D demosan 65W demasan chloronebe
Molecular Weight (g/mol):
207.05
Molecular Formula:
C8H8Cl2O2
InChi Key:
PFIADAMVCJPXSF-UHFFFAOYSA-N
SMILES:
COC1=CC(Cl)=C(OC)C=C1Cl
1
–
2
of
2
results
1
Chloroneb, TRC
CAS: 2675-77-6 Molecular Formula: C8 H8 Cl2 O2 Molecular Weight (g/mol): 207.05 Synonym: 1,4-Dichloro-2,5-dimethoxybenzene,2,5-Dichloro-1,4-dimethoxybenzene,Chloroneb,Demosan,Demosan 65W,FloPro D,NSC 151546,Soil Fungicide 1823 IUPAC Name: 1,4-dichloro-2,5-dimethoxybenzene SMILES: COc1cc(Cl)c(OC)cc1Cl